missing translation for 'onlineSavingsMsg'
Learn More
Learn More
His tag Rabbit anti-Tag, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10001-0-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The antibody reacts specifically with the 6xHis tag fused to recombit protein.
The 6x His tag is a synthetic oligo peptide consisting of 6 consecutive histidine residues (HHHHHH). The His tag is commonly expressed as a tag at either N-or C-terminal regions of recombit proteins to allow isolation or purification by immobilized metal affinity chromatography. Epitope tagging is a technique in which a known epitope is fused to a recombit protein using genetic engineering. By choosing a particular epitope and recombit protein combination, epitope tagging makes it possible to detect proteins for which no antibody is available. His epitope tagged proteins that contain a stretch of histidine residues at the carboxyl or amino terminus are used for purification of recombit proteins by means of metal chelate chromatography due to metal binding properties of histidine clusters on His tagged proteins. For example, the affinity of the histidine-tag motif to Ni2+ by chelation is strong and selective enough to enable purification of the protein to homogeneity by affinity chromatography on a Ni2+-NTA adsorbent.Specifications
His tag | |
Polyclonal | |
Unconjugated | |
6 his, 6x his,His tag, his tag | |
Antigen Affinity Chromatography | |
RUO | |
-20°C | |
Liquid | |
Tag |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Rabbit | |
150 μL | |
Primary | |
Antibody | |
IgG |