Learn More
Invitrogen™ HIPK1 (Myak) Recombinant Human Protein
HIPK1 phosphorylates homeodomain transcription factors.
4380.00 DKK - 57500.00 DKK
Specifications
Accession Number | NP_689909 |
---|---|
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 73.3kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10128252
|
Invitrogen™
PV4561 |
10μg |
4380.00 DKK
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15488663
|
Invitrogen™
PR8290A |
100μg |
13300.00 DKK
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15438733
|
Invitrogen™
PV4562 |
1mg |
57500.00 DKK
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
HIPK1 may also function as a co-repressor for homeodomain transcription factors.
Specifications
NP_689909 | |
73.3kDa | |
RUO - research use only | |
DYRK Kinase Family | |
Recombinant | |
Baculovirus |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
HIPK1 (Myak) | |
Human | |
GST Tag |