Få mere at vide
MedChemExpress Hinokitiol
Artikelnummer. 16458876
Encompass_Preferred
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
16458876 100 mg
1 options
Artikelnummer. 16458876 Leverandør MedChemExpress Leverandørnr. HYB2230
 Begrænset vare
Forventet forsendelse: 11-11-2026
MedChemExpress Hinokitiol is a component of essential oils isolated from Chymacyparis obtusa, reduces Nrf2 expression, and decreases DNMT1 and UHRF1 mRNA and protein expression, with anti-infective, anti-oxidative, and anti-tumor activities.
Tekniske data
Indhold og opbevaring Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molekylvægt (g/mol) 164.2 g/mol
Forsendelsestilstand Room temperature
Til brug med (applikation) Cancer-programmed cell death
CAS 499-44-5
Produkttype Natural Products
Renhed 98.0%
Opløselighed DMSO : 100 mg/mL (609.01 mM∣Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
Synonym β-Thujaplicin
Mål Keap1-Nrf2∣DNA Methyltransferase∣Virus Protease