Learn More
Fisherbrand™ Stainless Steel Hinged Covers for TI-H Ultrasonic Units
A safe and durable cover to complete your ultrasonic cleaning units. Fisherbrand™ Stainless Steel Hinged Covers for TI-H Ultrasonic Units have the added benefit of acoustic and thermal insulation.
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10713563
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Easy to clean
  • Strong and carefree
  • Clean-cut, smooth surfaces
  • Meets high safety standards
  • Acoustic and thermal insulation
  • Meets high user demands for hygiene
Specifications
Specifications
Lid
Ultrasonic Bath, TI-H20 16.8 L