Få mere at vide
Hamilton™ 1000μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips
Artikelnummer. 15293533
Skift visning
Pakningsstørrelse:
Pakke med 3840
Artikelnummer. unitSize
15293533 Pakke med 3840
1 options
Artikelnummer. 15293533 Leverandør Hamilton™ Leverandørnr. 235904_CORE_I
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Essential for precise pipetting. Hamilton™ 1000μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips provide enhanced pipetting safety and reliability with the CO-RE PerfectTouch system.
  • 95.1mm tip length
  • Color code label: white
  • Usable with 1000μL pipetting channels/CO-RE 96 Probe Head
  • Pressure-based Liquid Level Detection (pLLD) and capacitive (cLLD) with conductive (black) version
  • Process safety offered by Monitored Air Displacement and Total Aspiration and Dispense Monitoring
Tekniske data
Farve Sort
Volumen (metrisk) 1000 μL
Sterilitet Ikke-steril