Få mere at vide
Hamilton™ 1000μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips
Beskrivelse
- 95.1mm tip length
- Color code label: white
- Usable with 1000μL pipetting channels/CO-RE 96 Probe Head
- Pressure-based Liquid Level Detection (pLLD) and capacitive (cLLD) with conductive (black) version
- Process safety offered by Monitored Air Displacement and Total Aspiration and Dispense Monitoring
Tekniske data
Tekniske data
| Farve | Sort |
| Volumen (metrisk) | 1000 μL |
| Sterilitet | Ikke-steril |