Learn More
High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 149.2 mPa.s (60 °C), Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified high temperature viscosity standards are for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.
Brand:  Paragon Scientific S600S
Additional Details : Weight : 0.90000kg
Product Code. 16050380
  • 645.00 DKK / 500mL
Estimated Shipment: 07-06-2024
to see stock.
Description
Description
  • Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Kinematic (cSt), dynamic viscosity (mPa·s) and density (g/mL) given at all temperatures
  • Standard Pack Size 500mL
  • Fully traceable to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
Special Offers
Nominal values quoted. Certified Values will vary from batch to batch.