Learn More
Nabertherm™ High-Temperature Furnaces with Molybdenum Disilicide Heating Elements and Fiber Insulation
Due to their solid construction and compact stand-alone design, these high-temperature furnaces are perfect for processes in the laboratory where the highest precision is needed. Oustanding temperature uniformity and practical details set very high quality benchmarks.
Brand: Nabertherm™ HT0357VNA
190254.49 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Dual shell housing with fan cooling for low shell temperatures
- Heating from both sides via molybdenum disilicide heating elements
- High-quality fiber insulation backed by special insulation
- Side insulation constructed with tongue and groove blocks provides for low heat loss to the outside
- Long-life roof insulation with special suspension
- Chain-guided parallel swivel door for defined opening and closing of the door
- Vapor vent in the furnace roof with motorized exhaust air flaps, controlled via the extra function of the controller
- Heating elements switched via thyristors
Specifications
P570 | |
1750°C | |
300 x 200 x 260 mm | |
400 V |
16 L | |
Molybdenum Disilicide Heating Elements and Fiber Insulation | |
690 x 820 x 1860 mm | |
12500 W |