Learn More
Thermo Scientific™ High Temperature Ferrule Replacement Pk. for Exactive GC-Orbitrap GC-MS System
Product Code. 16834316 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Inside:
0.4 mm (Inner), 3.2 mm (Outer)
Unit Size:
Pack of 5
Product Code. Diameter (Metric) Inside Unit Size
16834316 0.4 mm (Inner), 3.2 mm (Outer) Pack of 5
1 options
Product Code. 16834316 Supplier Thermo Scientific™ Supplier No. 290VT221
Forventet forsendelse: 10-12-2026
Includes
Replacement Ferrules only
Allows for leak-free, high temperature transferline operation on Orbitrap-GC/MS with the ease-of-use of Vespel ferrules.
Use the Thermo Scientific™ GC-Orbitrap™ High Temperature Nut and Ferrule Kit to obtain leak-free results at high temperature transferline settings on your gas chromatography (GC) Orbitrap. The high-temperature SCP-5000 Vespel ferrules (included) allow continuous operating transferline temperatures of at least 320°C without ferrule deformation or breakdown (common with standard graphite/Vespel ferrules above 280°C), preventing leaks from forming over an extended period of time. The included spring loaded nut ensures a close fit between nut, ferrule and transferline, further preventing leaks from forming.
High-temperature SCP-5000 Vespel ferrules with 0.4mm ID can be used with most 0.1–0.25mm ID gas chromatography columns. Ferrules must be used in conjunction with High Temperature Adjustable Transferline Nut for GC-Orbitrap (included in kit). Nut and ferrules are for use with the Thermo Scientific™ Q Exactive™ GC Orbitrap™ GC-MS/MS system and the Thermo Scientific™ Exactive™ GC Orbitrap™ GC-MS system.
TRUSTED_SUSTAINABILITY
Specifications
Type High Temperature Ferrule Replacement Pk.
Description High Temperature Ferrule Replacement Pk.
Length (Metric) 2.7 mm
For Use With (Equipment) Thermo Scientific Q Exactive GC Orbitrap GC-MS/MS System and Thermo Scientific Exactive GC Orbitrap GC-MS System
Material Vespel
Quantity Pack of 5
Diameter (Metric) Inside 0.4 mm (Inner), 3.2 mm (Outer)