missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HFE Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99531UV
Additional Details : Weight : 0.00970kg
Description
HFE Polyclonal specifically detects HFE in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HFE | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
dJ221C16.10.1, hemochromatosis, hereditary hemochromatosis protein, hereditary hemochromatosis protein HLA-H, HH, high Fe, HLAH, HLA-HHFE1, MGC103790, MHC class I-like protein HFE, MVCD7 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse HFE. | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
3077 | |
Store at 4°C in the dark. | |
IgG |