missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
HEY1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
4670.00 DKK
Tekniske data
| Antigen | HEY1 |
|---|---|
| Applikationer | Immunocytochemistry, Immunofluorescence |
| Klassifikation | Polyclonal |
| Konjugeret | Unconjugated |
| Værtsarter | Rabbit |
Beskrivelse
HEY1 Polyclonal specifically detects HEY1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Tekniske data
| HEY1 | |
| Polyclonal | |
| Rabbit | |
| DNA Repair, DNA replication Transcription Translation and Splicing, Neuroscience, Transcription Factors and Regulators | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 23462 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MKRAHPEYSSSDSELDETIEVEKESADENGNLSSALGSMSPTTSSQILARKR | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| basic helix-loop-helix protein OAF1, BHLHb31, Cardiovascular helix-loop-helix factor 2, CHF-2, CHF2hHRT1, Class B basic helix-loop-helix protein 31, Hairy and enhancer of split-related protein 1, hairy/enhancer-of-split related with YRPW motif 1, hairy/enhancer-of-split related with YRPW motif protein 1, Hairy-related transcription factor 1, HERP2HES-related repressor protein 1, HESR-1, HESR1MGC1274, HES-related repressor protein 2, HRT1, HRT-1OAF1 | |
| HEY1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel