Learn More
Abcam Hexokinase Activity Assay Kit (Fluorometric)
Product Code. 32120065 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
Product Code. Quantity Unit Size
32120065 100 Tests 100 tests
1 options
Product Code. 32120065 Supplier Abcam Supplier No. ab211103100Test
 Restricted Item
Forventet forsendelse: 25-11-2026
Measure Hexokinase 1 activity in Cell/Tissue Extracts with Hexokinase Activity Assay Kit AB211103. Sensitivity = 2 μU. Cited in 2 publications. Use with Microplate reader.
Measure Hexokinase 1 activity in Cell/Tissue Extracts with Hexokinase Activity Assay Kit AB211103. Sensitivity = 2 μU. Cited in 2 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests
Regulatory Status RUO