Få mere at vide
Hexestrol-d6, TRC
Artikelnummer. 30405069
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30405069 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405069 Leverandør Toronto Research Chemicals Leverandørnr. TRCH2953021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexestrol hexane-1,1,1,6,6,6-d6
CAS 1215476-12-2
Molekylær formel C18 2H6 H16 O2
Mængde 1 mg
Synonym 4-[1,1,1,6,6,6-Hexadeuterio-4-(4-hydroxyphenyl)hexan-3-yl]phenol, 3,4-Di(p-hydroxyphenyl)-n-hexane-1,1,1,6,6,6-d6, Hexestrol-d6, 3,4-Bis(4-hydroxyphenyl)hexane-1,1,1,6,6,6-d6, Hexestrol-D6 (hexane-1,1,1,6,6,6-D6)
InChI formel InChI=1S/C18H22O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h5-12,17-20H,3-4H2,1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])CC(C(CC([2H])([2H])[2H])c1ccc(O)cc1)c2ccc(O)cc2
IUPAC navn 4-[1,1,1,6,6,6-hexadeuterio-4-(4-hydroxyphenyl)hexan-3-yl]phenol
Molekylvægt (g/mol) 276.4
Formel vægt 276.1996