Få mere at vide
Hexazinone-d6 (dimethyl-d6-amino), CDN
Artikelnummer. 30313243
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313243 0.01 g
30310854 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313243 Leverandør CDN Leverandørnr. CDND71460.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale HexazinoneD6(N,N-dimethylD6)
CAS 1219804-22-4
Molekylær formel C12 2H6 H14 N4 O2
Mængde 0.01 g
Synonym Hexazinone D6 (N,N-dimethyl D6), 6-[Bis(trideuteriomethyl)amino]-3-cyclohexyl-1-methyl-1,3,5-triazine-2,4-dione
InChI formel InChI=1 S/C12H20N4O2/c1-14(2)10-13-11(17)16(12(18)15(10)3)9-7-5-4-6-8-9/h9H,4-8H2,1-3H3/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])N(C1=NC(=O)N(C2CCCCC2)C(=O)N1C)C([2 H])([2 H])[2 H]
IUPAC navn 6-[bis(trideuteriomethyl)amino]-3-cyclohexyl-1-methyl-1,3,5-triazine-2,4-dione
Molekylvægt (g/mol) 258.35
Formel vægt 258.1963 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity