missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Hexazinone 1000 μg/mL in Toluene, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Hexazinone |
| CAS | 51235-04-2 |
| Analyt- eller komponentnavne | Hexazinone |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C12 H20 N4 O2 |
| Koncentration | 1000 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Velpar |
| InChI formel | InChI=1S/C12H20N4O2/c1-14(2)10-13-11(17)16(12(18)15(10)3)9-7-5-4-6-8-9/h9H,4-8H2,1-3H3 |
| SMIL | CN(C)C1=NC(=O)N(C2CCCCC2)C(=O)N1C |
| Vis mere |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?