Få mere at vide
Hexanoic Acid-d5, TRC
Artikelnummer. 30413349
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30413349 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413349 Leverandør Toronto Research Chemicals Leverandørnr. TRCP23777350MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexanoic Acid-d5
CAS 123167-40-8
Molekylær formel C6H7D5O2
Mængde 50 mg
Synonym Hexanoic-5,5,6,6,6-d5 Acid
InChI formel InChI=1S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8)/i1D3,2D2
SMIL OC(CCCC([2H])([2H])C([2H])([2H])[2H])=O
IUPAC navn hexanoic-5,5,6,6,6-d5 acid
Molekylvægt (g/mol) 121.19
Formel vægt 121.12