Få mere at vide
Hexanoic-5,5-d2 Acid, TRC
Artikelnummer. 30403172
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30403172 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403172 Leverandør Toronto Research Chemicals Leverandørnr. TRCH29215110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexanoic-5,5-D2 Acid
CAS 55320-68-8
Molekylær formel C6H10D2O2
Mængde 10 mg
Synonym Hexanoic-5,5-d2 acid (9CI), 5,5-Dideuteriohexanoic acid, Hexanoic acid-d2, Hexanoic-5,5-D2 acid, Caproic-Δ-d2 acid
InChI formel InChI=1S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8)/i2D2
SMIL CC([2H])([2H])CCCC(O)=O
IUPAC navn hexanoic-5,5-d2 acid
Molekylvægt (g/mol) 118.17
Formel vægt 118.1