Få mere at vide
Hexanoic-4,4,5,5,6,6,6-d7 Acid, CDN
Artikelnummer. 30325865
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30325865 0.25 g -
30318444 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325865 Leverandør CDN Leverandørnr. CDND73310.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexanoic-4,4,5,5,6,6,6-D7 Acid
CAS 1276197-47-7
Molekylær formel C6H5D7O2
Mængde 0.25 g
Synonym Hexanoic-4,4,5,5,6,6,6-d7 acid (ACI), 4,4,5,5,6,6,6-Heptadeuteriohexanoic acid, Hexanoic acid-d7, Hexanoic-4,4,5,5,6,6,6-D7 acid, Caproic-γ,delta;,ε-d7 acid
InChI formel InChI=1 S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8)/i1D3,2D2,3D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CCC(O)=O
IUPAC navn hexanoic-4,4,5,5,6,6,6-d7 acid
Molekylvægt (g/mol) 123.2
Formel vægt 123.13 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity