Få mere at vide
Hexanenitrile-6,6,6-d3, CDN
Artikelnummer. 30314552
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30314552 0.5 g 0.5g
30321312 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314552 Leverandør CDN Leverandørnr. CDND69310.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexanenitrile-6,6,6-D3
CAS 1219805-03-4
Molekylær formel C6 D3 H8 N
Mængde 0.5 g
Synonym Hexanenitrile-6,6,6-D3, 1-Cyanopentane-5,5,5-d3
InChI formel InChI=1 S/C6H11N/c1-2-3-4-5-6-7/h2-5H2,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])CCCCC#N
IUPAC navn 6,6,6-trideuteriohexanenitrile
Molekylvægt (g/mol) 100.1767
Formel vægt 100.108 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity