Få mere at vide
Hexanedioyl-d8 Chloride, CDN
Artikelnummer. 30308879
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30308879 0.25 g -
30319374 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30308879 Leverandør CDN Leverandørnr. CDND38560.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexanedioyl Dichloride-D8
CAS 284487-62-3
Molekylær formel C6 D8 Cl2 O2
Mængde 0.25 g
Synonym Hexanedioyl-d8 dichloride (9 CI), Hexanedioyl-d8 chloride, Hexanedioyl dichloride-D8, Adipoyl chloride-d8, Hexanedioic dichloride-d8, Adipic acid dichloride-d8
InChI formel InChI=1 S/C6H8Cl2O2/c7-5(9)3-1-2-4-6(8)10/h1-4H2/i1D2,2D2,3D2,4D2
SMIL [2 H]C([2 H])(C(=O)Cl)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)Cl
IUPAC navn 2,2,3,3,4,4,5,5-octadeuteriohexanedioyl dichloride
Molekylvægt (g/mol) 191.0818
Formel vægt 190.0403 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity