Få mere at vide
Hexanedioic-d8 Acid, TRC
Artikelnummer. 30321643
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321643 10 mg
30309645 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321643 Leverandør Toronto Research Chemicals Leverandørnr. TRCA29159110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Adipic Acid-D8
CAS 52089-65-3
Molekylær formel C6H2D8O4
Mængde 10 mg
Synonym Hexanedioic-2,2,3,3,4,4,5,5-D8 Acid, Adipic-D8 Acid, Hexanedioic-D8 Acid
InChI formel InChI=1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)/i1D2,2D2,3D2,4D2/hD2
SMIL OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(O)=O)=O
IUPAC navn dideuterio 2,2,3,3,4,4,5,5-octadeuteriohexanedioate
Molekylvægt (g/mol) 154.19