Få mere at vide
Hexanedioic Acid-1-13C, CDN
Artikelnummer. 30319159
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30319159 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319159 Leverandør CDN Leverandørnr. CDNC50401G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexanedioic Acid-1-13 C
CAS 2708283-72-9
Molekylær formel 13 C C5 H10 O4
Mængde 1 g
Synonym Hexanedioic-1-13 C acid, Adipic-1-13 C acid, Hexanedioic acid-1-13 C, Hexanedioic acid-13 C, Adipic Acid-13C1
InChI formel InChI=1 S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)/i5+1
SMIL O[13 C](=O)CCCCC(=O)O
IUPAC navn (613 C)hexanedioic acid
Molekylvægt (g/mol) 147.1339
Formel vægt 147.0613 g/mol
Procent renhed 99 atom % 13 C, min 98% Chemical Purity