missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexane, CHROMASOLV™, for HPLC, ≥95%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
6 x 1 L
Packaging:
Glass Bottle
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Melting Point | -95°C (literature) |
Absorbance | max. 0.02 at 240 nm, max. 0.01 at 250 nm, max. 0.01 at 400 nm, max. 1.0 at 195 nm, max. 0.30 at 210 nm, max. 0.10 at 220 nm, max. 0.05 at 230 nm |
CAS Min % | ≥90.0000% |
CAS Max % | ≤100.0000% |
Linear Formula | CH3(CH2)4CH3 |
UN Number | UN1208 |
Beilstein | 1730733 |
Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
Formula Weight | 86.18 |
Percent Purity | >95% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'