Learn More
Hexane-1-sulfonic acid sodium salt, LiChropur™ EMD-Millipore
Product Code. 15835890
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass
Unit Size:
25g
Product Code. Quantity Packaging Unit Size
15835890 25 g Glass 25g
1 options
Product Code. 15835890 Supplier Supelco Supplier No. 1183050025
 Restricted Item
Forventet forsendelse: 18-11-2026
Ion pair reagents for analytical HPLC LiChropur
TRUSTED_SUSTAINABILITY
Specifications
CAS 2832-45-3
Packaging Glass
Molecular Formula C6H13NaO3S
MDL Number MFCD00007542
Quantity 25 g
Synonym sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
InChI Key QWSZRRAAFHGKCH-UHFFFAOYSA-M
SMILES [Na+].CCCCCCS([O-])(=O)=O
IUPAC Name sodium hexane-1-sulfonate
Molecular Weight (g/mol) 188.22
PubChem CID 23677630
Formula Weight 188.22
Grade Not Available
Physical Form Powder
Chemical Name or Material Hexane-1-sulfonic acid sodium salt