Få mere at vide
Hexamidine Dihydrochloride, TRC
Artikelnummer. 30403926
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30403926 10 mg
30393838 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30403926 Leverandør Toronto Research Chemicals Leverandørnr. TRCH29420510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexamidine Dihydrochloride
CAS 50357-46-5
Molekylær formel C20H28Cl2N4O2
Mængde 10 mg
Synonym 4,4'-[1,6-Hexanediylbis(oxy)]bisbenzenecarboximidamide Hydrochloride, 1,6-Bis(4-amidinophenoxy)hexane, 4,4-Diamidino-1,6-diphenoxyhexane Hydrochloride, 4,4'-(Hexamethylenedioxy)dibenzamidine Hydrochloride, Desmodine Hydrochloride
InChI formel InChI=1S/C20H26N4O2.2ClH/c21-19(22)15-5-9-17(10-6-15)25-13-3-1-2-4-14-26-18-11-7-16(8-12-18)20(23)24;;/h5-12H,1-4,13-14H2,(H3,21,22)(H3,23,24);2*1H
SMIL Cl.Cl.NC(=N)c1ccc(OCCCCCCOc2ccc(cc2)C(=N)N)cc1
IUPAC navn 4-[6-(4-carbamimidoylphenoxy)hexoxy]benzenecarboximidamide;dihydrochloride
Molekylvægt (g/mol) 427.37
Formel vægt 426.1589