Få mere at vide
Hexamethyltriamidophosphate, TRC
Artikelnummer. 30008838
Skift visning
Mængde:
5 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008838 5 g
30012827 250 g
30005674 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008838 Leverandør Toronto Research Chemicals Leverandørnr. TRCH2943255G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 680-31-9
Molekylær formel C6 H18 N3 O P
Molekylvægt (g/mol) 179.2
Synonym HMPA, Hexamethylphosphoric triamide
IUPAC navn N-[bis(dimethylamino)phosphoryl]-N-methylmethanamine
SMIL CN(C)P(=O)(N(C)C)N(C)C
Tekniske data
Kemisk navn eller materiale Hexamethylphosphoramide (Hexametapol)
Mængde 5 g
InChI formel InChI=1S/C6H18N3OP/c1-7(2)11(10,8(3)4)9(5)6/h1-6H3
Formel vægt 179.1187