missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylenetetramine, 99%, Thermo Scientific Chemicals

Catalog Number p-4509637
364.59 DKK valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
100-97-0
C6H12N4
140.19
VKYKSIONXSXAKP-UHFFFAOYSA-N
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
4101
CHEBI:6824
C1N2CN3CN1CN(C2)C3
100-97-0
C6H12N4
140.19
VKYKSIONXSXAKP-UHFFFAOYSA-N
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
4101
CHEBI:6824
C1N2CN3CN1CN(C2)C3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 260.0°C
Color White to Yellow
Density 1.3300g/mL
pH 7 to 10 (1% aq. soln.)
Flash Point 250°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic Bottle
Quantity 1 kg
Beilstein 01,583
Fieser 01,427; 02,208; 04,243; 09,234
Merck Index 15,6038
Specific Gravity 1.33
Solubility Information Solubility in water: 895g/L (20°C). Other solubilities: soluble in chloroform: 13.4% (20°C), soluble in ethanol: 2.89% (20°C), soluble in acetone: 0.65% (20°C), insoluble in ether: 0.06% (20°C), soluble in methanol: 7.25% (20°C)
Sublimation Point 263.0 °C
Formula Weight 140.19
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material Hexamethylenetetramine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hexamethylenetetramine
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'