Få mere at vide
Hexamethyl-d18-disiloxane, TRC
Artikelnummer. 30408387
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30408387 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408387 Leverandør Toronto Research Chemicals Leverandørnr. TRCH29421625MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexamethyl-d18-disiloxane
CAS 19108-55-5
Molekylær formel C6 D18 O Si2
Mængde 25 mg
Synonym Disiloxane, 1,1,1,3,3,3-hexa(methyl-d3)- (ACI), 1,1,1,3,3,3-Hexa(methyl-d3)disiloxane (ACI), Disiloxane, hexa(methyl-d3)- (8CI, 9CI), Hexamethyl-d18-disiloxane, Hexamethyldisiloxane-d18, Hexamethyldisiloxane-D18
InChI formel InChI=1S/C6H18OSi2/c1-8(2,3)7-9(4,5)6/h1-6H3/i1D3,2D3,3D3,4D3,5D3,6D3
SMIL [2H]C([2H])([2H])[Si](O[Si](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn tris(trideuteriomethyl)-[tris(trideuteriomethyl)silyloxy]silane
Molekylvægt (g/mol) 180.4884
Formel vægt 180.2026