Få mere at vide
Hexamethonium Tartrate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Hexamethonium Tartrate |
| CAS | 2079-78-9 |
| Analyt- eller komponentnavne | Hexamethonium Tartrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C12 H30 N2 . 2 C4 H5 O6 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1,6-Hexanediaminium, N,N,N,N',N',N'-hexamethyl-, salt with (2R,3R)-2,3-dihydroxybutanedioic acid (1:2) (9CI), 1,6-Hexanediaminium, N,N,N,N',N',N'-hexamethyl-, salt with [R-(R*,R*)]-2,3-dihydroxybutanedioic acid (1:2), Ammonium, hexamethylenebis[trimethyl-, tartrate (1:2) (8CI), Hexamethylenebis[trimethylammonium hydrogen tartrate] (6CI,7CI), Butanedioic acid, 2,3-dihydroxy- (2R,3R)-, ion(1-), N,N,N,N',N',N'-hexamethyl-1,6-hexanediaminium (2:1) (9CI), Butanedioic acid, 2,3-dihydroxy-[R-(R*,R*)]-, ion(1-), N,N,N,N',N',N'-hexamethyl-1,6-hexanediaminium (2:1), Tartaric acid, ion(1-), hexamethylenebis[trimethylammonium] (2:1) (8CI), Esonium, Esotensyl, Hexamethonium bitartrate, Hexamethonium tartrate, Hexamethylenebistrimethylammonium di(hydrogen tartrate), Vegolysen-T |
| InChI formel | InChI=1S/C12H30N2.2C4H6O6/c1-13(2,3)11-9-7-8-10-12-14(4,5)6;2*5-1(3(7)8)2(6)4(9)10/h7-12H2,1-6H3;2*1-2,5-6H,(H,7,8)(H,9,10)/q+2;;/p-2/t;2*1-,2-/m.11/s1 |
| Løsningstype | Neat |
| SMIL | C[N+](C)(C)CCCCCC[N+](C)(C)C.O[C@H]([C@@H](O)C(=O)[O-])C(=O)O.O[C@H]([C@@H](O)C(=O)[O-])C(=O)O |
| Vis mere |