Få mere at vide
Hexahydro-1,3-isobenzofurandione-d8, TRC
Artikelnummer. 30407097
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30407097 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407097 Leverandør Toronto Research Chemicals Leverandørnr. TRCH29402225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexahydro-1,3-isobenzofurandione-d8
CAS 89614-24-4
Molekylær formel C8H2D8O3
Mængde 25 mg
Synonym 1,2-Cyclohexanedicarboxylic Anhydride-d8, 1,2-Cyclohexanedicarboxylic Acid Anhydride-d8, Aradur HY 925-d8, Araldite HT 904-d8, Araldite HT 907-d8, Araldite HY 907-d8, Araldite HY 925-d8, Araldite Hardener HY 925-d8, Cyclohexane-1,2-dicarboxylic Acid Anhydride-d8, Cyclohexanedicarboxylic Anhydride-d8, EP 400B-d8, Epilox H 11-01-d8, HHPA-d8, HT 907-d8, HY 925-d8, Hexahydro-1,3-isobenzofurandione-d8, Hexahydro-2-benzofuran-1,3-dione-d8, Hexahydrophthalic Acid Anhydride-d8, Hexahydrophthalic Anhydride-d8, Lekutherm Hardener H-d8, MH 700E-d8, NSC 8622-d8, Rikacid HH-d8, Rikacid HH-A-d8, Rikacid HHPA-d8, Rikacid MH 700E-d8, Rutadur AG-d8, Rutapox HX-d8
InChI formel InChI=1S/C8H10O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h5-6H,1-4H2/i1D2,2D2,3D2,4D2
SMIL O=C1OC(C2C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C21)=O
IUPAC navn hexahydroisobenzofuran-1,3-dione-4,4,5,5,6,6,7,7-d8
Molekylvægt (g/mol) 162.21
Formel vægt 162.11