Få mere at vide
Hexadecanoic-7,7,8,8-d4 Acid, CDN
Artikelnummer. 30314956
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30314956 0.1 g 0.1g
30310833 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314956 Leverandør CDN Leverandørnr. CDND18710.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexadecanoic-7,7,8,8-D4 Acid
CAS 75736-49-1
Molekylær formel C16 D4 H28 O2
Mængde 0.1 g
Synonym Hexadecanoic-7,7,8,8-d4 acid (9 CI, ACI), Hexadecanoic-7,7,8,8-D4 Acid, Palmitic acid-7,7,8,8-d4, [7,7,8,8-d4]Hexadecanoic acid, [7,7,8,8-d4]Palmitic acid, Hexadecanoic Acid-7,7,8,8-d4
InChI formel InChI=1 S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i9D2,10D2
SMIL [2 H]C([2 H])(CCCCCCCC)C([2 H])([2 H])CCCCCC(=O)O
IUPAC navn 7,7,8,8-tetradeuteriohexadecanoic acid
Molekylvægt (g/mol) 260.4487
Formel vægt 260.2653 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity