Få mere at vide
Hexadecanoic-3,3-d2 Acid, CDN
Artikelnummer. 30328418
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328418 0.05 g
30327127 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328418 Leverandør CDN Leverandørnr. CDND24160.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexadecanoic-3,3-D2 Acid
CAS 83293-32-7
Molekylær formel C16 D2 H30 O2
Mængde 0.05 g
Synonym Hexadecanoic-3,3-d2 acid (9 CI), 3,3-D2-Palmitic acid, Hexadecanoic-3,3-D2 Acid, Palmitic acid-3,3-d2, [3,3-2H2]Hexadecanoic acid, [3,3-2H2]Palmitic acid, Hexadecanoic Acid-3,3-d2
InChI formel InChI=1 S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i14D2
SMIL [2 H]C([2 H])(CCCCCCCCCCCCC)CC(=O)O
IUPAC navn 3,3-dideuteriohexadecanoic acid
Molekylvægt (g/mol) 258.4364
Formel vægt 258.2528 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity