Få mere at vide
Hexadecanoic-2,2-d2 Acid, CDN
Artikelnummer. 30316767
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30316767 1 g -
30326687 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316767 Leverandør CDN Leverandørnr. CDND16541G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Hexadecanoic-2,2-D2 Acid
CAS 62689-96-7
Molekylær formel C16 D2 H30 O2
Mængde 1 g
Synonym Hexadecanoic-2,2-d2 acid (9 CI, ACI), 2,2-D2-Palmitic acid, 2,2-d2-Palmitic acid, Palmitic acid-2,2-2H2, α-Dideuterohexadecanoic acid, [2,2-2H2]Hexadecanoic acid, Hexadecanoic-2,2-D2 acid
InChI formel InChI=1 S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i15D2
SMIL [2 H]C([2 H])(CCCCCCCCCCCCCC)C(=O)O
IUPAC navn 2,2-dideuteriohexadecanoic acid
Molekylvægt (g/mol) 258.4364
Formel vægt 258.2528 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity