Få mere at vide
Hexadecanoic-16,16,16-d3 Acid, CDN
Artikelnummer. 30322197
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322197 0.5 g
30321054 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322197 Leverandør CDN Leverandørnr. CDND16550.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Palmitic Acid-d3
CAS 75736-53-7
Molekylær formel C16 D3 H29 O2
Mængde 0.5 g
Synonym Hexadecanoic-16,16,16-d3 acid (9 CI, ACI), [16,16,16-2H3]Hexadecanoic acid, Hexadecanoic-16,16,16-d3 acid, Hexadecanoic acid-16,16,16-d3, Palmitic-16,16,16-d3 acid, Palmitic-ω-d3 acid
InChI formel InChI=1 S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i1D3
SMIL [2 H]C([2 H])([2 H])CCCCCCCCCCCCCCC(=O)O
IUPAC navn 16,16,16-trideuteriohexadecanoic acid
Molekylvægt (g/mol) 259.44
Formel vægt 259.2591 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity