Få mere at vide
Hexadecanedioic Acid Mono-L-carnitine Ester Chloride, TRC
Artikelnummer. 30405442
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30405442 25 mg
30408663 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30405442 Leverandør Toronto Research Chemicals Leverandørnr. TRCH29097025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 42150-38-9
Molekylær formel C23H44ClNO6
Molekylvægt (g/mol) 466.05
Synonym (R)-3-Carboxy-2-[(15-carboxy-1-oxopentadecyl)oxy]-N,N,N-trimethyl-1-propanaminium Chloride, (R)-3-((15-Carboxypentadecanoyl)oxy)-4-(trimethylammonio)butanoate Chloride
IUPAC navn [(2R)-3-carboxy-2-(15-carboxypentadecanoyloxy)propyl]-trimethylazanium;chloride
SMIL [Cl-].C[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CCCCCCCCCCCCCCC(=O)O
Tekniske data
Kemisk navn eller materiale Hexadecanedioic Acid Mono-L-carnitine Ester Chloride
Mængde 25 mg
InChI formel InChI=1S/C23H43NO6.ClH/c1-24(2,3)19-20(18-22(27)28)30-23(29)17-15-13-11-9-7-5-4-6-8-10-12-14-16-21(25)26;/h20H,4-19H2,1-3H3,(H-,25,26,27,28);1H/t20-;/m1./s1
Formel vægt 465.2857