Få mere at vide
Hexaconazole, MedChemExpress
Artikelnummer. 16481986
Encompass_Preferred
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16481986 500 mg
16458046 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16481986 Leverandør Medchem Express Leverandørnr. HYA0278
MedChemExpress Hexaconazole is a systemic fungicide used for the control of many fungi particularly Ascomycetes and Basidiomycetes.
Hexaconazole is a systemic fungicide used for the control of many fungi particularly Ascomycetes and Basidiomycetes.
Tekniske data
Kemisk navn eller materiale Hexaconazole
Fysisk form Solid
CAS 79983-71-4
Farve White
Mængde 500 mg
Renhedsgrad noter Research
Molekylær formel C14H17Cl2N3O
Synonym (-)-Hexaconazol
Opløselighedsinformation DMSO : ≥ 100 mg/mL (318.26 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL ClC1=CC(Cl)=C(C(CCCC)(O)CN2N=CN=C2)C=C1
Molekylvægt (g/mol) 314.21
Formel vægt 314.21
Procent renhed 98.12%