Få mere at vide
Hexachlorobenzene-13C6, TRC
Artikelnummer. 30006997
Skift visning
Mængde:
1 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006997 10 mg
30007515 1 mg
30005414 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006997 Leverandør Toronto Research Chemicals Leverandørnr. TRCH29081710MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 93952-14-8
Molekylær formel 13C6 Cl6
Molekylvægt (g/mol) 290.74
Synonym Benzene-13C6, hexachloro- (9CI), Hexachlorobenzene 13C6
IUPAC navn 1,2,3,4,5,6-hexachloro(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene
SMIL Cl[13c]1[13c](Cl)[13c](Cl)[13c](Cl)[13c](Cl)[13c]1Cl
Tekniske data
Kemisk navn eller materiale Hexachlorobenzene 13C6
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vægt 287.8332
Procent renhed >95%