missing translation for 'onlineSavingsMsg'
Learn More

Hexachloro-1,3-butadiene-13C4, CDN

Product Code. 30324025
Change view
Click to view available options
Mængde:
0.01 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30324025 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324025 Supplier CDN Supplier No. CDNC24080.01G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Hexachloro-1,3-butadiene13C4
CAS 93951-70-3
Molekylær formel 13C4 Cl6
Mængde 0.01 g
Synonym 1,3-Butadiene-13C4, 1,1,2,3,4,4-hexachloro- (9 CI), Hexachlorobutadiene-13C4, Hexachloro-1,3-butadiene 13C4, 1,1,2,3,4,4-Hexachloro-1,3-butadiene-13C4
InChI formel InChI=1 S/C4Cl6/c5-1(3(7)8)2(6)4(9)10/i1+1,2+1,3+1,4+1
SMIL Cl[13 C](=[13 C](Cl)[13 C](=[13 C](Cl)Cl)Cl)Cl
IUPAC navn 1,1,2,3,4,4-hexachloro(1,2,3,4-13C4)buta-1,3-diene
Molekylvægt (g/mol) 264.7314
Formel vægt 261.8265 g/mol
Procent renhed 99 atom % 13 C, min 97% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.