missing translation for 'onlineSavingsMsg'
Få mere at vide

Hex-5-yn-1-amine, TRC

Product Code. 30397272
Change view
Click to view available options
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30397272 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397272 Supplier Toronto Research Chemicals Supplier No. TRCH290150500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Hex-5-yn-1-amine
CAS 15252-45-6
Molekylær formel C6 H11 N
Mængde 500 mg
Synonym 5-Hexynylamine, 1-Aminohex-5-yne, 6-Amino-1-hexyne
InChI formel InChI=1S/C6H11N/c1-2-3-4-5-6-7/h1H,3-7H2
SMIL NCCCCC#C
IUPAC navn hex-5-yn-1-amine
Molekylvægt (g/mol) 97.158
Formel vægt 97.089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.