Få mere at vide
Heteronium Bromide, LoGiCal
Artikelnummer. 30463062
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463062 10 mg
1 options
Artikelnummer. 30463062 Leverandør LoGiCal Leverandørnr. LGCFOR3090.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Heteronium Bromide
CAS 7247-57-6
Analyt- eller komponentnavne Heteronium Bromide
Forsendelsestilstand Room Temperature
Molekylær formel C18 H22 N O3 S . Br
Certificeringer/compliance ISO 9001
Synonym Pyrrolidinium, 3-[[2-hydroxy-2-phenyl-2-(2-thienyl)acetyl]oxy]-1,1-dimethyl-, bromide (1:1), Pyrrolidinium, 3-[(hydroxyphenyl-2-thienylacetyl)oxy]-1,1-dimethyl-, bromide (9CI), 31814, Heteronium bromide, Hetrum Bromide, 3-Hydroxy-1,1-dimethylpyrrolidinium bromide, alpha-phenyl-2-thiopheneglycolate (7CI), Pyrrolidinium, 3-hydroxy-1,1-dimethyl-, bromide, alpha-phenyl-2-thiopheneglycolate (8CI), (+/-)-Heteronium bromide
InChI formel InChI=1S/C18H22NO3S.BrH/c1-19(2)11-10-15(13-19)22-17(20)18(21,16-9-6-12-23-16)14-7-4-3-5-8-14;/h3-9,12,15,21H,10-11,13H2,1-2H3;1H/q+1;/p-1
Løsningstype Neat
SMIL [Br-].C[N+]1(C)CCC(C1)OC(=O)C(O)(c2ccccc2)c3cccs3
IUPAC navn (1,1-dimethylpyrrolidin-1-ium-3-yl) 2-hydroxy-2-phenyl-2-thiophen-2-ylacetate;bromide
Industry Type Clinical Testing
Molekylvægt (g/mol) 412.34
Mængde 10 mg
Formel vægt 411.0504 g/mol