Få mere at vide
Thermo Scientific™ HERAfreeze™ HERAfreeze™ HDE Series -40°C Ultra-Low Temperature Freezers
Artikelnummer. 16470915 Shop alle Thermo Scientific produkter
Elektriske krav:
230 V, 50 Hz
Pakningsstørrelse:
Hver
Artikelnummer. 16470915
Brand: Thermo Scientific™ HERAfreeze™ HDE40040FV
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Thermo Scientific™ HERAfreeze™ HDE Series -40°C ultra-low temperature freezers feature reduced energy usage, a simplified touchscreen interface, three capacities, and an ergonomic cabinet design.
Thermo Scientific™ HERAfreeze™ HDE Series -40°C ultra-low temperature freezers, powered by H-drive, feature three upright models, maximizing storage capacity from 40,000 up to 60,000 2mL vials. The HDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every lab.
Performance
  • Powered by H-drive's hydrocarbon technology, these systems save up to 50% in energy usage and heat rejection over conventional refrigerant ultra-low freezers1
  • A quiet sound output below 53 dBA, enables the freezer to come out of the hallway and into the lab
  • Meets ±5°C validation protocols at -40°C with strong peak variation and steady-state temperature variation
  • Natural hydrocarbon refrigerants reduce system operating stresses promoting outstanding reliability and worry-free sample protection
User Friendly
  • The H-drive Information Center (HIC) combines easy to read temperature with simple capacitive touch buttons in a 5.6 inch display
  • Three capacities (400, 500 and 600 2-inch box) to accommodate a variety of storage needs and lab footprint specifications
  • All models contain four inner doors allowing for easy sample access without exposing greater freezer content to ambient conditions
  • Improved customer convenience with new ergonomically designed handle, high contrast eye-level display, and comfortable easy-access shelf locations.
  • Compatible with most racking systems including a full range of Thermo Scientific™ racks for boxes, microplates, Thermo Scientific™ Matrix™ and Nunc CryoBank™ tubes
  • Security features including alarm settings and padlock-compatible doors
Sustainability
  • Manufactured in an award-winning, zero-waste to landfill facility
  • Lower HVAC costs as a result of less heat emission into the environment
  • Natural refrigerants and water-blown foam insulation, making the Series compliant with the U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), the European Union's F-Gas and other sustainability standards
Footnotes:
  1. Internal data comparing 3.9 kWh/day to DXF400 = 8.0 kWh/day. Data on file.
  2. Typical freezer test data based on internal testing in ambient temperature at 20°C; Typical standard validation protocol of ±5°C.
TRUSTED_SUSTAINABILITY
Tekniske data
Oversigt HERAfreeze HDE Series -40°C Ultra-Low Freezers, 400-Box Capacity, 230V, 50Hz
Kapacitet (engelsk) 19.4 cu. ft.
Kapacitet (metrisk) 549 L
Temperaturområde -10°C to -40°C
Dør stil Outer (1), Inner (5)
Antal døre 5
Isolering Vacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
Indre Stainless Steel
Hylder 4
Kapacitet (engelsk) hylde 165 lb.
Kapacitet (metrisk) hylde 75 kg
Certificeringer/compliance CE
Stil Upright
Vise HIC 5.6 in. 7-segment display with capacitive touch buttons
Produktlinje HERAfreeze
garanti Warranty varies by country, please contact us for details
Afrimning Manual
Dimensioner (DxBxH) Udvendig 38.5 x 28.3 x 78 in. (97.7 x 71.9 x 198.1cm)
Mål (D x B x H) Indvendig 28.3 x 23.1 x 51.2 in. (71.9 x 58.8 x 130.1cm)
Overvågningsmuligheder Alarm, Recorder
Sætpunktssikkerhed Yes
Forbindelse Remote Alarm Connections, 4-20mA, RS485, and Dry Contacts
Smart View-kompatibilitet Smart-Vue Compatible
Strømstyrke 3.2/10
Stiktype EU
Spænding 230 V
Elektriske krav 230 V, 50 Hz
Frekvens 50 Hz
Hætteglaskapacitet 40,000 x 2mL
Kompressor H-drive
Kølemiddel R290
Forsendelsesvægt (engelsk) 642 lb.
Forsendelsesvægt (metrisk) 292kg
Vægt (engelsk) 550 lb.
Vægt (metrisk) 249 kg
Holder 2 in. Boxes (400)