Få mere at vide
Heptanoic-2,2-d2 Acid, CDN
Artikelnummer. 30320859
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320859 0.5 g
30316990 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320859 Leverandør CDN Leverandørnr. CDND53350.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Heptanoic Acid-2,2-D2
CAS 64118-38-3
Molekylær formel C7 D2 H12 O2
Mængde 0.5 g
Synonym Heptanoic-2,2-d2 acid (9 CI), Heptanoic Acid-2,2-D2, Heptanoic acid-d2, Enanthic acid-d2
InChI formel InChI=1 S/C7H14O2/c1-2-3-4-5-6-7(8)9/h2-6H2,1H3,(H,8,9)/i6D2
SMIL [2 H]C([2 H])(CCCCC)C(=O)O
IUPAC navn 2,2-dideuterioheptanoic acid
Molekylvægt (g/mol) 132.1972
Formel vægt 132.1119 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity