missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane, Extra Pure, Fraction, SLR, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
Description
Heptane, Extra Pure, Fraction, SLR
Specifications
Packaging | Amber glass bottle |
Quantity | 1 L |
Color | Colorless |
IUPAC Name | Petroleum ether |
Formula Weight | 100.2g/mol |
Vapor Pressure | 48mbar at 20°C |
Viscosity | 0.4 mPaS at 20°C |
Vapor Density | 3.5 |
Melting Point | -90°C |
Boiling Point | 90°C to 100°C |
Show More |
Safety and Handling
Product Identifier
- Heptane
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'