Få mere at vide
Heptanal-d14, CDN
Artikelnummer. 30313186
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313186 0.25 g
30311023 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313186 Leverandør CDN Leverandørnr. CDND73020.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Heptanal-d14
CAS 1246819-97-5
Molekylær formel C7 D14 O
Mængde 0.25 g
Synonym 1,2,2,3,3,4,4,5,5,6,6,7,7,7-Tetradecadeuterioheptan-1-one, Heptanal-d14, Heptanal-D14, Heptanal-1,2,2,3,3,4,4,5,5,6,6,7,7,7-d14, 1-Heptaldehyde-d14, Enanthal-d14, Enanthaldehyde-d14, Enanthic Aldehyde-d14, Enanthole-d14, Heptaldehyde-d14, Heptanaldehyde-d14, Heptyl Aldehyde-d14, Oenanthal-d14, Oenanthaldehyde-d14, Oenanthic Aldehyde-d14, Oenanthol-d14, n-Heptaldehyde-d14, n-Heptanal-d14, n-Heptylaldehyde-d14
InChI formel InChI=1 S/C7H14O/c1-2-3-4-5-6-7-8/h7H,2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7 D
SMIL [2 H]C(=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,2,2,3,3,4,4,5,5,6,6,7,7,7-tetradecadeuterioheptan-1-one
Molekylvægt (g/mol) 128.2717
Formel vægt 128.1923 g/mol
Procent renhed 98 atom % D, min 96% Chemical Purity