Få mere at vide
Heptaminol Hydrochloride, TRC
Artikelnummer. 30404281
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30404281 50 mg
30395588 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30404281 Leverandør Toronto Research Chemicals Leverandørnr. TRCH54616050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Heptaminol hydrochloride
CAS 543-15-7
Molekylær formel C8 H19 N O . Cl H
Mængde 50 mg
Synonym Heptaminol Hydrochloride, 6-amino-2-methylheptan-2-ol hydrochloride
InChI formel InChI=1S/C8H19NO.ClH/c1-7(9)5-4-6-8(2,3)10;/h7,10H,4-6,9H2,1-3H3;1H
SMIL Cl.CC(N)CCCC(C)(C)O
IUPAC navn 6-amino-2-methylheptan-2-ol;hydrochloride
Molekylvægt (g/mol) 181.7
Formel vægt 181.1233