Få mere at vide
Heptaminol Hydrochloride, LoGiCal
Artikelnummer. 30462485
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462485 10 mg
1 options
Artikelnummer. 30462485 Leverandør LoGiCal Leverandørnr. LGCFOR0653.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Heptaminol hydrochloride
CAS 543-15-7
Analyt- eller komponentnavne Heptaminol hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C8 H19 N O . Cl H
Certificeringer/compliance ISO 9001
Synonym Heptaminol Hydrochloride, 6-amino-2-methylheptan-2-ol hydrochloride
InChI formel InChI=1S/C8H19NO.ClH/c1-7(9)5-4-6-8(2,3)10;/h7,10H,4-6,9H2,1-3H3;1H
Løsningstype Neat
SMIL Cl.CC(N)CCCC(C)(C)O
IUPAC navn 6-amino-2-methylheptan-2-ol;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 181.7
Mængde 10 mg
Formel vægt 181.1233 g/mol