Få mere at vide
Heptadecanoic Acid (D33, 98%), TRC
Artikelnummer. 30405889
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30405889 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405889 Leverandør Toronto Research Chemicals Leverandørnr. TRCH998798100MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Heptadecanoic Acid (D33)
CAS 352431-41-5
Molekylær formel C17 D33 H O2
Mængde 100 mg
Synonym Heptadecanoic-d33 acid (9CI), Heptadecanoic Acid-D33, n-Heptadecylic acid-d33, Margaric acid-d33
InChI formel InChI=1S/C17H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19/h2-16H2,1H3,(H,18,19)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)O
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,17-tritriacontadeuterioheptadecanoic acid
Molekylvægt (g/mol) 303.654
Formel vægt 303.463