Learn More
Hepatitis D Virus Large delta Protein Recombinant Rabbit Monoclonal Antibody (HL1051), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA542362
Additional Details : Weight : 0.01000kg
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Hepatitis D Virus Large delta Protein Recombinant Monoclonal antibody specifically detects Hepatitis D Virus Large delta Protein in Virus samples. It is validated for Western Blot.Specifications
Hepatitis D Virus Large delta Protein | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
Western Blot | |
HL1051 | |
Unconjugated | |
HDV | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Hepatitis D virus Large delta protein (Hepatitis delta virus genotype I (isolate D380). The exact sequence is proprietary. | |
100 μL | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |