missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heparin Lithium salt, MedChemExpress

Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Heparin Lithium salt is an anticoagulant which binds reversibly to antithrombin III (ATIII). Heparin Lithium salt significantly inhibits exosome-cell interactions.
Specifications
Specifications
Chemical Name or Material | Heparin Lithium salt |
Physical Form | Solid |
CAS | 9045-22-1 |
Color | White |
Quantity | 100 mg |
Purity Grade Notes | Research |
Molecular Formula | (C14H25NO20S3)n.xLi |
Solubility Information | H2O : 100 mg/mL (Need ultrasonic) |
SMILES | O[C@H]1[C@H](O[C@H]2[C@H](OS(O)(=O)=O)[C@@H](O)[C@H](OC)[C@H](C(O)=O)O2)[C@@H](COS(O)(=O)=O)O[C@H](OC)[C@@H]1NS(O)(=O)=O.[n].[.xLi] |
Grade | Research |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Heparin Lithium salt, MedChemExpress
Spot an opportunity for improvement?Share a Content Correction