Få mere at vide
Hemiacetorphan, TRC
Artikelnummer. 30137853
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137853 250 mg
30006832 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137853 Leverandør Toronto Research Chemicals Leverandørnr. TRCH235105250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 124735-06-4
Molekylær formel C14 H17 N O4 S
Molekylvægt (g/mol) 295.35
Synonym N-[2-[(Acetylthio)methyl]-1-oxo-3-phenylpropyl]glycine, Glycine, N-[2-[(acetylthio)methyl]-1-oxo-3-phenylpropyl]-, (+/-)-, Hemiacetorphan, S-Acetylthiorphan, [[(2RS)-2-[(Acetylsulfanyl)methyl]-3-phenylpropanoyl]amino]acetic Acid
IUPAC navn 2-[[2-(acetylsulfanylmethyl)-3-phenylpropanoyl]amino]acetic acid
SMIL CC(=O)SCC(Cc1ccccc1)C(=O)NCC(=O)O
Tekniske data
Kemisk navn eller materiale Hemiacetorphan
Mængde 250 mg
InChI formel InChI=1S/C14H17NO4S/c1-10(16)20-9-12(14(19)15-8-13(17)18)7-11-5-3-2-4-6-11/h2-6,12H,7-9H2,1H3,(H,15,19)(H,17,18)
Formel vægt 295.0878