missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ HELIOS Polyclonal Antibody
Beskrivelse
This antibody predicted to react with Monkey, Sheep, Cat.
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predominantly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.
Tekniske data
Tekniske data
| Antigen | HELIOS |
| Applikationer | Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Koncentration | 0.5 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4 |
| Gene | IKZF2 |
| Genadgangsnr. | Q9UKS7 |
| Gene Alias | A730095J18Rik; ANF1A2; Helios; IKAROS family zinc finger 2; IKAROS family zinc finger 2 (Helios); ikaros family zinc finger protein 2; Ikzf2; Zfpn1a2; zinc finger DNA binding protein Helios; zinc finger protein Helios; zinc finger protein, subfamily 1A, 2 (Helios); ZNF1A2; Znfn1a2 |
| Gensymboler | IKZF2 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?